Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals FSB Buffer

Catalog No. p-7082223
Change view
Click to view available options
Quantity:
125 mL
250 mL
Catalog No. Quantity
AAJ60262EQE 125 mL
AAJ60262AK 250 mL
2 options

Catalog No. AAJ60262EQE

Supplier: Thermo Scientific Chemicals J60262EQE

Only null left
Add to Cart

For frozen storage of competent cells

For frozen storage of competent cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer
Color Colorless
For Use With (Application) For frozen storage of competent cells
Physical Form Liquid
Quantity 125 mL
Synonym Frozen storage buffer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.