Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSBA Antibody (FSBA 2F5/5), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB310414 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310414 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310414 Supplier Novus Biologicals Supplier No. NB1002656MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FSBA Monoclonal specifically detects FSBA in Non-species specific samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen FSBA
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone FSBA 2F5/5
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias 5'-(4-(fluorosulfonyl)benzoyl)adenosine, 5'-(4-fluorosulfonylbenzoyl)adenosine, 5'-FSBA, p-fluorosulfonylbenzoyl-5'-adenosine
Host Species Mouse
Immunogen Fluorylsulponylbenzoyladenosine (FSBA)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.