Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSH beta Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB090134 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB090134 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB090134 Supplier Novus Biologicals Supplier No. NBP299911B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FSH beta Polyclonal specifically detects FSH beta in Human samples. It is validated for ELISA.

Specifications

Antigen FSH beta
Applications ELISA
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias follicle stimulating hormone, beta polypeptide, Follicle-stimulating hormone beta subunit, Follitropin beta chain, follitropin subunit beta, follitropin, beta chain, FSH-B, FSH-beta
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant FSH beta (NP_000501.1; Met1-Glu129).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2488
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.