Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSHR Antibody (626717), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB174287 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174287 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174287 Supplier Novus Biologicals Supplier No. FAB65591E
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FSHR Monoclonal antibody specifically detects FSHR in Human samples. It is validated for Flow Cytometry

Specifications

Antigen FSHR
Applications Flow Cytometry
Classification Monoclonal
Clone 626717
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias follicle stimulating hormone receptor, follicle-stimulating hormone receptor, Follitropin receptor, FSH receptor, FSH-R, LGR1FSHRO, MGC141667, MGC141668, ODG1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human FSH R, Accession # M65085
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2492
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.