Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSHR Antibody (FSHR/1400), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP254329G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254329G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP254329G Supplier Novus Biologicals Supplier No. NBP254329G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FSHR Monoclonal specifically detects FSHR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FSHR
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FSHR/1400
Conjugate DyLight 488
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias follicle stimulating hormone receptor, follicle-stimulating hormone receptor, Follitropin receptor, FSH receptor, FSH-R, LGR1FSHRO, MGC141667, MGC141668, ODG1
Gene Symbols FSHR
Host Species Mouse
Immunogen Recombinant human FSHR protein (Uniprot: P23945)
Purification Method Protein A or G purified
Quantity 100 μL
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2492
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.