Learn More
FSHR Polyclonal Antibody

Description
FSHR Polyclonal Antibody for IHC (P)
Specifications
Specifications
| Antigen | FSHR |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | FSHR |
| Gene Accession No. | P20395, P23945, P35376, P35379, P47799, P49059 |
| Gene Alias | follicle stimulating hormone receptor; follicle-stimulating hormone receptor; Follitropin receptor; FSH receptor; Fshr; FSH-R; FSHR1; FSHRO; LGR1; ODG1 |
| Gene Symbols | FSHR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.