Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FTO Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB11060935
Description
FTO Polyclonal specifically detects FTO in Human, Mouse, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoblotting.Specifications
FTO | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
Rabbit | |
60 kDa | |
0.1 mL | |
Lipid and Metabolism, Neuroscience | |
79068 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunoblot | |
1.1 mg/mL | |
Western Blot 0.5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 28205605), Immunoblotting reported in scientific literature (PMID 22019092) | |
Q9C0B1 | |
FTO | |
A synthetic peptide within the C-terminal region (residues 400-505) of the human FTO protein. [Swiss-Prot# Q9C0B1] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Chicken, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction