Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTO Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$207.00

Specifications

Antigen FTO
Concentration 1.1 mg/mL
Dilution Western Blot 0.5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 28205605), Immunoblotting reported in scientific literature (PMID 22019092)
Applications Western Blot, Immunohistochemistry, Immunoblot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11060935T
SDP
View Documents
Novus Biologicals
NB11060935SS
0.025 mL
Each for $207.00
Only null left
Add to Cart
 
NB11060935
SDP
View Documents
Novus Biologicals
NB11060935
0.1 mL N/A N/A N/A
Description

Description

FTO Polyclonal specifically detects FTO in Human, Mouse, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoblotting.
Specifications

Specifications

FTO
Western Blot 0.5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 28205605), Immunoblotting reported in scientific literature (PMID 22019092)
Polyclonal
Rabbit
Lipid and Metabolism, Neuroscience
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149
FTO
IgG
Affinity Purified
60 kDa
1.1 mg/mL
Western Blot, Immunohistochemistry, Immunoblot
Unconjugated
RUO
Human, Mouse, Chicken, Primate
Q9C0B1
79068
A synthetic peptide within the C-terminal region (residues 400-505) of the human FTO protein. [Swiss-Prot# Q9C0B1]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.