Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTO Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB339084 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339084 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339084 Supplier Novus Biologicals Supplier No. NB11060935PCP

Rabbit Polyclonal Antibody

FTO Polyclonal antibody specifically detects FTO in Human, Mouse, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoblot

Specifications

Antigen FTO
Applications Western Blot, Immunohistochemistry, Immunoblot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunoblotting
Formulation PBS
Gene Alias alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149
Host Species Rabbit
Immunogen A synthetic peptide within the C-terminal region (residues 400-505) of the human FTO protein. [Swiss-Prot# Q9C0B1]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 79068
Target Species Human, Mouse, Chicken, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.