Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FTSJ3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
Antigen | FTSJ3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17929620
![]() |
Novus Biologicals
NBP17929620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179296
![]() |
Novus Biologicals
NBP179296 |
100 μL |
Each for $499.50
|
|
|||||
Description
FTSJ3 Polyclonal specifically detects FTSJ3 in Rat samples. It is validated for Western Blot.Specifications
FTSJ3 | |
Polyclonal | |
Rabbit | |
NP_001012014 | |
117246 | |
Synthetic peptide directed towards the C terminal of human Ftsj3The immunogen for this antibody is Ftsj3. Peptide sequence QVTYVVAKKGVGRKVRRPAGVRGHFKVVDSRMKKDQRAQRKEQKRNHRRK. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EC 2.1.1, EC 2.1.1.-, EPCS3, FLJ20062, FtsJ homolog 3 (E. coli), Protein ftsJ homolog 3, putative rRNA methyltransferase 3, rRNA (uridine-2'-O-)-methyltransferase 3 | |
FTSJ3 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title