Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTSJ3 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB327787 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327787 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327787 Supplier Novus Biologicals Supplier No. NBP305948PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FTSJ3 Polyclonal antibody specifically detects FTSJ3 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen FTSJ3
Applications Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias EC 2.1.1, EC 2.1.1.-, EPCS3, FLJ20062, FtsJ homolog 3 (E. coli), Protein ftsJ homolog 3, putative rRNA methyltransferase 3, rRNA (uridine-2'-O-)-methyltransferase 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FTSJ3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 117246
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.