Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FtsZ (GTPase), Rabbit anti-Bacteria, Polyclonal, Millipore Sigma™
Rabbit Polyclonal Antibody
$192.34 - $482.80
Specifications
Antigen | FtsZ (GTPase) |
---|---|
Dilution | Western Blotting Analysis: A representative lot detected FtsZ (GTPase) in Western Blotting applications (Lucet, I., et. al. (2000). EMBO J. 19(7):1467-75; Bisson-Filho, A.W., et. al. (2017). Science. 355(6326):739-743).Immunofluorescence Analysis: A representative lot detected FtsZ (GTPase) in Immunofluorescence applications (Daniel, R.A., et. al. (2000). Mol Microbiol. 35(2):299-311). |
Applications | Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABS220025UL
|
MilliporeSigma
ABS220025UL |
25 μL |
Each for $192.34
|
|
|||||
ABS2200100
|
MilliporeSigma
ABS2200100UL |
100 μL |
Each for $482.80
|
|
|||||
Description
Anti-FtsZ (GTPase), Cat. No. ABS2200, is a rabbit polyclonal antibody that detects Cell division protein FtsZ and has been tested for use in Immunofluorescence and Western Blotting.
Cell division protein FtsZ (UniProt: P17865; also known as Cell Division FtsZ GTPase, FtsZ) is encoded by the FtsZ (also known as BSU15290) gene (Gene ID: 935971) in Bacillus subtilis. FtsZ is homodimeric prokaryotic homologue of tubulin with GTPase activity. It is found in nearly all bacteria. It polymerizes in the presence of GTP to form higher-ordered polymers. It contains three GTP-binding sites that are located in the N-terminal region. The less conserved C-terminal domain contains residues that are important for GTP hydrolysis. FtsZ is shown to be essential for bacterial cell division and in association with FtzA (FtsAZ) it forms a contractile ring structure (Z ring) at the site of cell division. The regulation of the ring assembly controls the timing and the location of cell division. One of the major functions of FtsZ is to recruit other cell division proteins to the septum to produce a new cell wall between the dividing cells. The FtsZ treadmilling rate controls the rate of peptidoglycan synthesis and cell division. During bacterial cell division FtsZ co-localizes with SpoIIE, a dual function protein that contributes to the switch form medial to polar cell division during sporulation. The anti-FtsZ blot has also been used as control for sample loading. (Ref.: Bisson-Filho, AW., et al., (2017). Science 355(6326); 739-743; Feucht, A., and Jeffery Errington, J (2005). Microbiology 151(6); 2053-2064; Lucet, I., et al. (2000). EMBO J. 19(7); 1467-1475). Lytvynenko, I., et al. (2019). Cell. 178(1); 76-90)Specifications
FtsZ (GTPase) | |
Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Signaling | |
Rabbit polyclonal antiserum with 0.05% sodium azide. | |
NP_389412 | |
Full length purified FtsZ from Bacillus subtilis. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Western Blotting Analysis: A representative lot detected FtsZ (GTPase) in Western Blotting applications (Lucet, I., et. al. (2000). EMBO J. 19(7):1467-75; Bisson-Filho, A.W., et. al. (2017). Science. 355(6326):739-743).Immunofluorescence Analysis: A representative lot detected FtsZ (GTPase) in Immunofluorescence applications (Daniel, R.A., et. al. (2000). Mol Microbiol. 35(2):299-311). | |
Polyclonal | |
Serum | |
RUO | |
Bacteria | |
Cell division protein FtsZ;Cell Division FtsZ GTPase | |
FtsZ;BSU15290 | |
IgG | |
Unpurified | |
This rabbit polyclonal antibody detects Cell Division Protein FTsZ in Bacillus subtilis. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title