Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUBI/MNSF beta/FAU Rabbit anti-Human, DyLight 594, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB101894 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB101894 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB101894 Supplier Novus Biologicals Supplier No. NBP297414DL594

Rabbit Polyclonal Antibody

FUBI/MNSF beta/FAU Polyclonal specifically detects FUBI/MNSF beta/FAU in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FUBI/MNSF beta/FAU
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias 40S ribosomal protein S30, asr1, FAU1, FAU-encoded ubiquitin-like protein, FBR-MuSV-associated ubiquitously expressed, Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed, Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed(fox derived), FLJ22986, Fub1, Fubi, MNSFbeta, monoclonal nonspecific suppressor factor beta, ribosomal protein S30, RPS30, ubiquitin-like protein fubi and ribosomal protein S30, ubiquitin-like-S30 fusion protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FUBI/MNSF beta/FAU fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2197
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.