Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUBP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216542 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216542 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216542 Supplier Novus Biologicals Supplier No. NBP216542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FUBP1 Polyclonal specifically detects FUBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen FUBP1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7), 20% Glycerol with 0.025% Proclin 300
Gene Accession No. Q96AE4
Gene Alias far upstream element (FUSE) binding protein 1, far upstream element binding protein, far upstream element-binding protein 1, FBPDNA helicase V, FUBP, FUSE-binding protein 1, hDH V
Gene Symbols FUBP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FUBP1. The exact sequence is proprietary.
Molecular Weight of Antigen 68 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 8880
Test Specificity Human. Expected to cross react based on sequence identity: Mouse (99%), Rat (99%), Xenopus laevis (91%), Bovine (99%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.