Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fucosyltransferase 3/FUT3 Antibody, Novus Biologicals™
SDP

Catalog No. NB395279 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395279 25 μL
NBP254732 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB395279 Supplier Novus Biologicals Supplier No. NBP25473225UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fucosyltransferase 3/FUT3 Polyclonal specifically detects Fucosyltransferase 3/FUT3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Blood Group Lewis b
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias alpha-(1,3/1,4)-fucosyltransferase, Blood group Lewis alpha-4-fucosyltransferase, CD174, EC 2.4.1, EC 2.4.1.65, FT3B, Fucosyltransferase 3, fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood group), Fucosyltransferase III, FucT-III, galactoside 3(4)-L-fucosyltransferase, LEfucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood groupincluded), Les, Lewis FT, MGC131739
Gene Symbols FUT3
Host Species Rabbit
Immunogen This antibody was developed against a Recombinant Protein corresponding to amino acids:KDLARYLQELDKDHARYLSYFRWRETLRPRSFSWAL
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2525
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.