Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fucosyltransferase 8/FUT8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | Fucosyltransferase 8/FUT8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17986920
![]() |
Novus Biologicals
NBP17986920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179869
![]() |
Novus Biologicals
NBP179869 |
100 μL |
Each for $487.50
|
|
|||||
Description
Fucosyltransferase 8/FUT8 Polyclonal specifically detects Fucosyltransferase 8/FUT8 in Human samples. It is validated for Western Blot.Specifications
Fucosyltransferase 8/FUT8 | |
Polyclonal | |
Rabbit | |
NP_004471 | |
2530 | |
The immunogen for this antibody is FUT8. Peptide sequence LVQRRITYLQNPKDCSKAKKLVCNINKGCGYGCQLHHVVYCFMIAYGTQR. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
alpha-(1,6)-fucosyltransferase, alpha1-6FucT, EC 2.4.1.68, Fucosyltransferase 8, fucosyltransferase 8 (alpha (1,6) fucosyltransferase), GDP-fucose--glycoprotein fucosyltransferase, GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha1,6-fucosyltransferase, Glycoprotein 6-alpha-L-fucosyltransferase, MGC26465 | |
FUT8 | |
IgG | |
47 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title