Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fullerene, nanotube, multi-walled, 20 nm OD, 5-20 micron long

Catalog No. AA43839MF
Click to view available options
Quantity:
50 mg
250 mg

CAS: 308068-56-6 | CH4 | 16.043 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 308068-56-6
Molecular Formula CH4
Molecular Weight (g/mol) 16.043
MDL Number MFCD00133992
InChI Key VNWKTOKETHGBQD-UHFFFAOYSA-N
Synonym methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
PubChem CID 297
ChEBI CHEBI:16183
IUPAC Name methane
SMILES C

Specifications

Color Black
Mesh Size −270 Mesh
Quantity 50 mg
UN Number UN1325
Physical Form Powder
Chemical Name or Material Fullerene, Nanotube, multi-walled, 20nm OD, 5 to 20 micron long
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.