Learn More
Fullerene powder, 99.5% C{60}
Description
Fullerene powder is useful for biomedical applications such as the design of high-performance MRI contrast agents, x-ray imaging contrast agents and photodynamic therapy. It is also essential for single-walled carbon nanotubes with narrow diameter-distribution. It is used as light-activated antimicrobial agent. It also finds application in the field of nanotechnology, heat resistance and superconductivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Black-Brown |
| Density | 1.7g/mL |
| Boiling Point | 500°C to 600°C (sublimation) |
| Quantity | 5 g |
| UN Number | UN1325 |
| Percent Purity | 99.5% |
| Physical Form | Powder |
| Assay | C60 |
| Chemical Name or Material | Fullerene powder |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H228-H315-H319-H335 Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P210-P261-P280-P305+P351+P338-P405-P501a Keep away from heat/sparks/open flames/hot surfaces. No smoking. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S. |
| RTECSNumber | LS9252500 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.