Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumonisin Antibody (2A2), Novus Biologicals™
SDP

Catalog No. NBP221061 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221061 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221061 Supplier Novus Biologicals Supplier No. NBP221061
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fumonisin Monoclonal specifically detects Fumonisin in Non-species specific samples. It is validated for ELISA, Antibody Competition.

Specifications

Antigen Fumonisin
Applications ELISA, Competition Assay
Classification Monoclonal
Clone 2A2
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA 1:2000-1:10000, Antibody Competition
Gene Alias Fumonisin
Host Species Mouse
Immunogen Purified BSA-Fumonisin
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with Fusarium Fumonisin mycotoxin and B1 and B2 subgroups of Fumonisin.
Reconstitution Reconstitute with 0.2 ml distilled water.
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.