Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUR1 Antibody (OTI2D1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288197 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288197 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288197 Supplier Novus Biologicals Supplier No. NBP272399APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FUR1 Monoclonal antibody specifically detects FUR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen FUR1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2D1
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias DKFZp781E1243, EC 2.4.2, EC 2.4.2.9, FUR1, MGC23937, RP11-311P8.3, UMP pyrophosphorylase, UPP, UPRTase, uracil phosphoribosyltransferase (FUR1) homolog (S. cerevisiae), uracil phosphoribosyltransferase homolog
Host Species Mouse
Immunogen Full length human recombinant protein of human UPRT(NP_659489) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 139596
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.