Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furegrelate Sodium Salt, ≥99%, Thermo Scientific Chemicals

Catalog No. aaj61378lb0
Click to view available options
Quantity:
5 mg

Inhibitor of thrombaxane synthetase

Inhibitor of thrombaxane synthetase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85666-17-7
Molecular Formula C15H10NNaO3, C15H10NNaO3
Synonym U-63557A; Sodium furegrelate

Specifications

Quantity 5 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Inhibitor of thrombaxane synthetase
Formula Weight 275.20g/mol
Solubility Soluble in water at approximately 15mg/ml
Product Type Signal Transduction Reagent
Form Crystalline solid
InChI Key XBTIPIZROJAKOJ-UHFFFAOYSA-M
SMILES [Na+].[O-]C(=O)C1=CC2=CC(CC3=CC=CN=C3)=CC=C2O1
IUPAC Name sodium 5-[(pyridin-3-yl)methyl]-1-benzofuran-2-carboxylate
Molecular Weight (g/mol) 275.24
Assay ≥99%
Show More Show Less
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.