Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furin Antibody (1C3), Novus Biologicals™
SDP

Catalog No. NB297851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297851 100 μL
NB297850 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297851 Supplier Novus Biologicals Supplier No. NBP326563100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Furin Monoclonal antibody specifically detects Furin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen Furin
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 1C3
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Furin [UniProt P09958]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5045
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.