Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furin Antibody (222722), Biotin, Novus Biologicals™
SDP

Catalog No. NB297105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297105 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297105 Supplier Novus Biologicals Supplier No. FAB1503B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Furin Monoclonal specifically detects Furin in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunoprecipitation, Western Blot.

Specifications

Antigen Furin
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 222722
Conjugate Biotin
Dilution Western Blot, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Furin, Asp108-Glu715, Accession # NP_002560
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5045
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.