Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furin Antibody (MM0298-4G31), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP22321646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22321646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP22321646 Supplier Novus Biologicals Supplier No. NBP212321JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Furin Monoclonal specifically detects Furin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Furin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0298-4G31
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1
Gene Symbols FURIN
Host Species Mouse
Immunogen Human recombinant Furin
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Golgi Apparatus Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5045
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.