Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furin Antibody, Novus Biologicals™
SDP

Catalog No. NB1001903 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001903 Supplier Novus Biologicals Supplier No. NB1001903
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Furin Polyclonal specifically detects Furin in Canine, Hamster, Human, Mouse, Pig, Primate, Rat samples. It is validated for Block/Neutralize, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Furin
Applications Blocking Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 3 - 5 ug, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess
Formulation PBS, 1 mg/ml BSA with 0.05% Sodium Azide
Gene Alias Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1
Gene Symbols FURIN
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues R(780) G E R T A F I K D Q S A L(793) of human Furin.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5045
Test Specificity Detects Furin convertase from canine and mouse cells as well as transfected human Furin. This does not detect endogenous Furin from BSC-40, HeLa, J774A.1 BPAEC, or CHO cells nor from rat skeletal muscle, spleen, kidney, ovary, testes, heart, or brain tissues.
Target Species Canine, Hamster, Rat, Human, Mouse, Pig, Primate, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.