Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUS Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291839 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291839 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291839 Supplier Novus Biologicals Supplier No. NB100560APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FUS Polyclonal specifically detects FUS in Human, Mouse samples. It is validated for Immunoprecipitation (Negative), Western Blot.

Specifications

Antigen FUS
Applications Immunoprecipitation (Negative), Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation PBS
Gene Alias 75 kDa DNA-pairing protein, ALS6, amyotrophic lateral sclerosis 6, FUS1, fused in sarcoma, fusion (involved in t(12,16) in malignant liposarcoma), fusion gene in myxoid liposarcoma, fusion, derived from t(12,16) malignant liposarcoma, fus-like protein, heterogeneous nuclear ribonucleoprotein P2, HNRNPP2, Oncogene FUS, Oncogene TLS, POMp75, RNA-binding protein FUS, TLShnRNP-P2, Translocated in liposarcoma protein
Host Species Rabbit
Immunogen A synthetic peptide which maps to a region between residues 200 and 250 of human fusion (involved in t(12,16) in malignant liposarcoma) using the numbering given in SwissProt entry P35637 (GeneID 2521)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2521
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.