Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUS Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB092607 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092607 100 μL
NB092608 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092607 Supplier Novus Biologicals Supplier No. NBP303651100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FUS Polyclonal specifically detects FUS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen FUS
Applications Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias 75 kDa DNA-pairing protein, ALS6, amyotrophic lateral sclerosis 6, FUS1, fused in sarcoma, fusion (involved in t(12;16) in malignant liposarcoma), fusion gene in myxoid liposarcoma, fusion, derived from t(12;16) malignant liposarcoma, fus-like protein, heterogeneous nuclear ribonucleoprotein P2, HNRNPP2, Oncogene FUS, Oncogene TLS, POMp75, RNA-binding protein FUS, TLShnRNP-P2, Translocated in liposarcoma protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 297-526 of human FUS (NP_004951.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2521
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.