Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUS2 Antibody (2F4), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP141157Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141157Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141157Y Supplier Novus Biologicals Supplier No. NBP141157AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FUS2 Monoclonal specifically detects FUS2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FUS2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2F4
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Gene Accession No. NP_036323
Gene Alias FUS-2, FUS2EC 2.3.1.-, N-acetyltransferase 6, N-acetyltransferase 6 (GCN5-related), Protein fus-2, Protein fusion-2
Gene Symbols NAA80
Host Species Mouse
Immunogen Recombinant FUS2 (1-308aa) purified from E. coli
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 24142
Test Specificity This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human FUS2 protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.