Learn More
Cancer Diagnostics, Inc.™ Fusion Micro Scalpel
SDP
Catalog No. 22500460 Shop All Cancer Diagnostics Products
Click to view available options
Product Type:
Scalpel (Handle Only)
Scalpel Combo Pack
Turns microtome blades into a knife
  • Get more use out of your old microtome blades—simply secure blade into Fusion handle and use as a knife or scalpel
  • Single-use handle and low-profile blades in dispenser pack available separately
  • Combo pack includes 50 handles and 50 blades
Specifications
Product Type Scalpel Combo Pack
Quantity 50 Single Use Handles and 50 Low Profile Blades
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.