Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUSIP1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB050638 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB050638 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB050638 Supplier Novus Biologicals Supplier No. NBP297993G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FUSIP1 Polyclonal specifically detects FUSIP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FUSIP1
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 40 kDa SR-repressor protein, FLJ43846, FUS interacting protein (serine-arginine rich) 1, FUS-interacting protein (serine-arginine rich) 2, FUS-interacting serine-arginine-rich protein 1, FUSIP1DKFZp686H0644, FUSIP2, neural-salient SR protein, NSSR, serine/arginine-rich splicing factor 10, serine-arginine repressor protein (40 kDa), SFRS13, SFRS13A, Splicing factor SRp38, splicing factor, arginine/serine-rich 13, Splicing factor, arginine/serine-rich 13ATLS-associated protein with Ser-Arg repeats, SR splicing factor 10, SRp38, SRrp40TLS-associated protein with SR repeats, TASR1TLS-associated SR protein, TASR2FUS interacting protein (serine/arginine-rich) 1, TASRFLJ30749, TLS-associated protein TASR, TLS-associated serine-arginine protein, TLS-associated serine-arginine protein 1, TLS-associated serine-arginine protein 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FUSIP1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10772
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.