Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUSIP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB396314 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396314 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB396314 Supplier Novus Biologicals Supplier No. NBP24681525UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FUSIP1 Polyclonal antibody specifically detects FUSIP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen FUSIP1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2), 40% Glycerol
Gene Accession No. O75494
Gene Alias 40 kDa SR-repressor protein, FLJ43846, FUS interacting protein (serine-arginine rich) 1, FUS-interacting protein (serine-arginine rich) 2, FUS-interacting serine-arginine-rich protein 1, FUSIP1DKFZp686H0644, FUSIP2, neural-salient SR protein, NSSR, serine/arginine-rich splicing factor 10, serine-arginine repressor protein (40 kDa), SFRS13, SFRS13A, Splicing factor SRp38, splicing factor, arginine/serine-rich 13, Splicing factor, arginine/serine-rich 13ATLS-associated protein with Ser-Arg repeats, SR splicing factor 10, SRp38, SRrp40TLS-associated protein with SR repeats, TASR1TLS-associated SR protein, TASR2FUS interacting protein (serine/arginine-rich) 1, TASRFLJ30749, TLS-associated protein TASR, TLS-associated serine-arginine protein, TLS-associated serine-arginine protein 1, TLS-associated serine-arginine protein 2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: DRKTPNQMKAKEGRNVYSSSRYDDYDRYRRSRSR
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10772
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.