Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUT10 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB323738 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323738 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323738 Supplier Novus Biologicals Supplier No. NBP299776PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FUT10 Polyclonal antibody specifically detects FUT10 in Human samples. It is validated for ELISA

Specifications

Antigen FUT10
Applications ELISA
Classification Polyclonal
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias alpha 1,3-fucosyl transferase, alpha-(1,3)-fucosyltransferase 10, EC 2.4.1, EC 2.4.1.-, Fucosyltransferase 10, fucosyltransferase 10 (alpha (1,3) fucosyltransferase), Fucosyltransferase X, Fuc-TX, FucT-X, Galactoside 3-L-fucosyltransferase 10, MGC11141
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human FUT10 (Uniprot#: Q6P4F1-1; Leu 32-Asp 479)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84750
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.