Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUZ Antibody, Novus Biologicals™
SDP

Catalog No. NBP224750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224750 0.1 mg
NBP224750SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224750 Supplier Novus Biologicals Supplier No. NBP224750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FUZ Polyclonal specifically detects FUZ in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FUZ
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-6 ug/ml, Immunohistochemistry-Paraffin 15 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9BT04
Gene Alias FLJ22688, fuzzy homolog (Drosophila), FY, protein fuzzy homolog
Gene Symbols FUZ
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 338-382 of human FUZ was used as immunogen for the antibody, GenBank no NP_079405.2.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80199
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, chimpanzee, opossum, horse, rat, mouse, cow and dog and 94% homologous in chicken, Xenopus, zebrafish and Tetradon.
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.