Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXR/NR1H4 Antibody, Novus Biologicals™
SDP

Catalog No. NB400153 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400153 0.1 mL
NB400153SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400153 Supplier Novus Biologicals Supplier No. NB400153
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

FXR/NR1H4 Polyclonal specifically detects FXR/NR1H4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FXR/NR1H4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 2.4 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-4 ug/ml, Immunocytochemistry/Immunofluorescence 1:100
Formulation Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
Gene Alias bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14RXR-interacting protein 14
Gene Symbols NR1H4
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus of the human FXR protein. [UniProt# Q96RI1]
Molecular Weight of Antigen 66 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9971
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.