Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXR2 Antibody (1G2), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB30259A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB30259A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB30259A750 Supplier Novus Biologicals Supplier No. NB300259AF750

Mouse Monoclonal Antibody

FXR2 Monoclonal specifically detects FXR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FXR2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1G2
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FMR1L2, fragile X mental retardation syndrome-related protein 2, fragile X mental retardation, autosomal homolog 2, fragile X-mental retardation 1-like 2, FXR2P
Gene Symbols FXR2
Host Species Mouse
Immunogen Recombinant C-terminal fragment of human FXR2 (residues 414-658). [UniProtNo. P51116]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9513
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.