Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXR2 Antibody (1G2), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB300259UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300259UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300259UV Supplier Novus Biologicals Supplier No. NB300259UV

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

FXR2 Monoclonal specifically detects FXR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FXR2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1G2
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FMR1L2, fragile X mental retardation syndrome-related protein 2, fragile X mental retardation, autosomal homolog 2, fragile X-mental retardation 1-like 2, FXR2P
Gene Symbols FXR2
Host Species Mouse
Immunogen Recombinant C-terminal fragment of human FXR2 (residues 414-658). [UniProt# P51116]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cellular Markers, Chromatin Research, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9513
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.