Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXYD2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB043121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB043121 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB043121 Supplier Novus Biologicals Supplier No. NBP297415F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FXYD2 Polyclonal specifically detects FXYD2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FXYD2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ATP1C, ATP1G1ATPase, Na+/K+ transporting, gamma 1 polypeptide, FXYD domain containing ion transport regulator 2, FXYD domain-containing ion transport regulator 2, HOMG2, hypomagnesemia 2, renal, MGC12372, Na(+)/K(+) ATPase subunit gamma, Sodium pump gamma chain, sodium/potassium-transporting ATPase subunit gamma, Sodium-potassium-ATPase, gamma polypeptide
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FXYD2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 486
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.