Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXYD5/Dysadherin Antibody (CL14037), Novus Biologicals™
SDP

Catalog No. NB306000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306000 100 μL
NB305999 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB306000 Supplier Novus Biologicals Supplier No. NBP324585100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FXYD5/Dysadherin Monoclonal antibody specifically detects FXYD5/Dysadherin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen FXYD5/Dysadherin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL14037
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias DYSAD, dysadherin, FXYD domain containing ion transport regulator 5, FXYD domain-containing ion transport regulator 5, IWU1, KCT1, keratinocytes associated transmembrane protein 1, OIT2, PRO6241, RIC
Host Species Mouse
Immunogen This antibody was designed to target the recombinant protein FXYD5/Dysadherin, with the precise immunogen sequence being withheld for proprietary reasons.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 53827
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.