Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXYD5/Dysadherin Antibody (CL14037), Novus Biologicals™

Mouse Monoclonal Antibody
$342.50 - $534.50
Specifications
Antigen | FXYD5/Dysadherin |
---|---|
Clone | CL14037 |
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
FXYD5/Dysadherin Monoclonal antibody specifically detects FXYD5/Dysadherin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
FXYD5/Dysadherin | |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DYSAD, dysadherin, FXYD domain containing ion transport regulator 5, FXYD domain-containing ion transport regulator 5, IWU1, KCT1, keratinocytes associated transmembrane protein 1, OIT2, PRO6241, RIC | |
This antibody was designed to target the recombinant protein FXYD5/Dysadherin, with the precise immunogen sequence being withheld for proprietary reasons. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL14037 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Signal Transduction | |
PBS, pH 7.2, 40% glycerol | |
53827 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title