Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXYD5/Dysadherin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159138
Description
FXYD5/Dysadherin Polyclonal specifically detects FXYD5/Dysadherin in Human samples. It is validated for Western Blot.Specifications
FXYD5/Dysadherin | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
DYSAD, dysadherin, FXYD domain containing ion transport regulator 5, FXYD domain-containing ion transport regulator 5, IWU1, KCT1, keratinocytes associated transmembrane protein 1, OIT2, PRO6241, RIC | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q96DB9 | |
FXYD5 | |
Synthetic peptides corresponding to FXYD5(FXYD domain containing ion transport regulator 5) The peptide sequence was selected from the middle region of FXYD5. Peptide sequence SERPSPSTDVQTDPQTLKPSGFHEDDPFFYDEHTLRKRGLLVAAVLFITG. | |
100 μL | |
Signal Transduction | |
53827 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction