Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXYD5/Dysadherin Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen FXYD5/Dysadherin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159138
SDP
View Documents
Novus Biologicals
NBP159138
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

FXYD5/Dysadherin Polyclonal specifically detects FXYD5/Dysadherin in Human samples. It is validated for Western Blot.
Specifications

Specifications

FXYD5/Dysadherin
Polyclonal
Rabbit
Human
DYSAD, dysadherin, FXYD domain containing ion transport regulator 5, FXYD domain-containing ion transport regulator 5, IWU1, KCT1, keratinocytes associated transmembrane protein 1, OIT2, PRO6241, RIC
FXYD5
IgG
Western Blot
Unconjugated
RUO
Q96DB9
53827
Synthetic peptides corresponding to FXYD5(FXYD domain containing ion transport regulator 5) The peptide sequence was selected from the middle region of FXYD5. Peptide sequence SERPSPSTDVQTDPQTLKPSGFHEDDPFFYDEHTLRKRGLLVAAVLFITG.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.