Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FYB/ADAP/SLAP130 Antibody (460107), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB348109 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348109 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348109 Supplier Novus Biologicals Supplier No. FAB38571MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FYB/ADAP/SLAP130 Monoclonal antibody specifically detects FYB/ADAP/SLAP130 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen FYB/ADAP/SLAP130
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 460107
Conjugate mFluor Violet 610 SE
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ADAP, Adhesion and degranulation promoting adaptor protein, adhesion and degranulation-promoting adaptor protein, FYB-120/130SLAP-130FYN-binding protein (FYB-120/130), FYN binding protein, FYN-binding protein, FYN-T-binding protein, p120/p130, PRO0823, SLAP130, SLP-76-associated phosphoprotein
Host Species Mouse
Immunogen E. coli-derived recombinant human FYB/ADAP/SLAP130, Ala2-Gly274, Accession # O15117
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2533
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.