Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fyn Antibody (FYN-01), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB500517V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500517V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500517V Supplier Novus Biologicals Supplier No. NB500517V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fyn Monoclonal specifically detects Fyn in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Fyn
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FYN-01
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias EC 2.7.10, EC 2.7.10.2, FYN oncogene related to SRC, FGR, YES, MGC45350, OKT3-induced calcium influx regulator, p59-Fyn, protein-tyrosine kinase fyn, Proto-oncogene c-Fyn, Proto-oncogene Syn, proto-oncogene tyrosine-protein kinase fyn, SLKc-syn protooncogene, src/yes-related novel, Src-like kinase, SYN, tyrosine kinase p59fyn(T), tyrosine-protein kinase Fyn
Gene Symbols FYN
Host Species Mouse
Immunogen Bacterial expressed recombinant fragment, corresponding to amino acids 7-176 of Human Fyn.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2534
Test Specificity The reacts with p59fyn (Fyn), a non receptor protein tyrosine kinase (59 kDa),which belongs to the Src family of cytoplasmic tyrosine kinases.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.