Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fyn Antibody, Novus Biologicals™
SDP

Catalog No. NBP132067 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132067 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132067 Supplier Novus Biologicals Supplier No. NBP132067
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fyn Polyclonal specifically detects Fyn in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Fyn
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal
Gene Alias EC 2.7.10, EC 2.7.10.2, FYN oncogene related to SRC, FGR, YES, MGC45350, OKT3-induced calcium influx regulator, p59-Fyn, protein-tyrosine kinase fyn, Proto-oncogene c-Fyn, Proto-oncogene Syn, proto-oncogene tyrosine-protein kinase fyn, SLKc-syn protooncogene, src/yes-related novel, Src-like kinase, SYN, tyrosine kinase p59fyn(T), tyrosine-protein kinase Fyn
Gene Symbols FYN
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FYN. The exact sequence is proprietary.
Molecular Weight of Antigen 61 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2534
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.