Learn More
Bacharach™ Fyrite™ CO2 or O2 Analyzer Refill Kits
Keep your analyzer working with the Fyrite CO2 or O2 Analyzer Refill Kit.
| Catalog Number | Mfr. No. | Includes | Concentration Measurement Range | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Concentration Measurement Range | Price | Quantity & Availability | ||||
|
10-884-5
|
Bacharach™
11 7047 |
2 Bottles of fluid (20/60% CO2), top gasket screws, filtering material | 0 to 20% CO | N/A | N/A | N/A | |||
|
10-884-15
|
Bacharach™
11 7050 |
2 Bottles of fluid (21/60% O2), top gasket screws, filtering material | 0 to 20% CO | N/A | N/A | N/A | |||
|
10-884-6
|
Bacharach™
10 5100 |
3 Bottles of fluid (7.6% CO2) | 0 to 7.6% CO | N/A | N/A | N/A | |||
Description
- Include two bottles of fluid, top gasket screws, filtering material 10-884-6 consists of three bottles of fluid only
Specifications
| CO2 Analyzer Refill Kit |