Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FZR1/CDH1 Antibody (AR38.2), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP219018IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219018IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219018IR Supplier Novus Biologicals Supplier No. NBP219018IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FZR1/CDH1 Monoclonal specifically detects FZR1/CDH1 in Human, Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen FZR1/CDH1
Applications Western Blot, ELISA
Classification Monoclonal
Clone AR38.2
Conjugate DyLight 755
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CDC20C, CDH1, Cdh1/Hct1 homolog, CDH1CDC20-like 1b, fizzy/cell division cycle 20 related 1 (Drosophila), Fzr, FZR2, FZRCDC20-like protein 1, hCDH1, HCDHFYR, KIAA1242fizzy-related protein homolog
Gene Symbols FZR1
Host Species Mouse
Immunogen His-tagged recombinant FZR1/CDH1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 51343
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.