Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FZR1/CDH1 Antibody (DH01 (DCS-266)), Novus Biologicals™
SDP

Catalog No. NBP222103 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222103 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222103 Supplier Novus Biologicals Supplier No. NBP222103

Mouse Monoclonal Antibody

FZR1/CDH1 Monoclonal specifically detects FZR1/CDH1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen FZR1/CDH1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone DH01 (DCS-266)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunoprecipitation 2 ug/mg
Gene Accession No. Q9UM11
Gene Alias anti-CDC20-like protein 1 anti-CDH1 anti-Cdh1/Hct1 homolog anti-Fizzy-related protein homolog anti-FYR anti-Fzr anti-FZR1 anti-hCDH1 CDC20C, CDH1, Cdh1/Hct1 homolog, CDH1CDC20-like 1b, fizzy/cell division cycle 20 related 1 (Drosophila), FYR, Fzr, FZR2, FZRCDC20-like protein 1, hCDH1, HCDHFYR, KIAA1242, KIAA1242fizzy-related protein homolog
Gene Symbols FZR1
Host Species Mouse
Immunogen Recombinant human cdh1 protein
Purification Method Protein G purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51343
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.