Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ G 28UCM

Catalog No. p-7110827
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

FASN inhibitor

G 28UCM is a FASN inhibitor. Inhibits breast cancer cell proliferation in vitro (IC50 values are 21 - 79 μM), including cell lines resistant to anti-HER2 therapies. Blocks HER2 signaling and induces apoptosis. Inhibits growth of breast cancer xenografts in mice without causing anorexia.

Specifications

Chemical Name or Material 3-(3,4,5-Trihydroxybenzoyloxy)naphthalen-1-yl 3,4,5-trihydroxybenzoate
CAS 1094451-90-7
Quantity 10 mg
Target Fatty Acid Synthase Inhibitors
Molecular Formula C24H16O10
Formula Weight 464.38
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.