Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-CSF Antibody (243), Alexa Fluor™ 594
SDP

Catalog No. NB065852 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB065852 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB065852 Supplier Novus Biologicals Supplier No. NBP289191AF594

Rabbit Monoclonal Antibody

G-CSF Monoclonal specifically detects G-CSF in Human samples. It is validated for Sandwich ELISA Detection.

Specifications

Antigen G-CSF
Applications ELISA
Classification Monoclonal
Clone 243
Conjugate Alexa Fluor 594
Dilution Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human G-CSF (NP_757373.1; Ala30-Pro204).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 1440
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.