Learn More
Description
Specifications
Specifications
| Antigen | G-CSF |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 243 |
| Conjugate | DyLight 755 |
| Dilution | Sandwich ELISA Detection |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human G-CSF (NP_757373.1; Ala30-Pro204). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
